Learn More
Description
Host Species: Mouse
Clone: CT-1
Isotype: IgG1/κ
Species Reactivity: Human, Mouse and Rat.
Epitope: aa-GRVLTLPRS- within the C-terminus
Immunogen: A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor β-subunit
Molecular Weight: 95kDa
Positive Control: IM-9 lymphocyte cells
Cellular Localization: Membrane Ab-5 reacts with autophosphorylated and unoccupied receptors as well as the receptor-insulin complex. Ab-5 does not affect insulin-stimulated autophosphorylation.
Recommended for:
- Immunoprecipitation
- Affinity Purification (With peptide elution)
- ELISA (As capture antibody only)
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Insulin Receptor (beta-Subunit) Ab-5 |
| Applications | Immunoprecipitation |
| Classification | Monoclonal |
| Clone | CT-1 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor Beta-subunit |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.