Learn More
Description
Host Species: Mouse
Clone: CT-3
Isotype: IgG1
Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.
Epitope: C-terminal
Immunogen: Recombinant fragment including the C-terminal 100 amino acids of human insulin receptor
Molecular Weight: 95kDa
Positive Control: HEP-G-2, IM-9 lymphocyte cells; Placenta or brain
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Not suitable)
- Affinity Purification (With peptide elution)
- ELISA (As capture antibody only)
- Tyrosine Kinase Assay (Ab-mediated capture on microplates)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Insulin Receptor (beta-Subunit) Ab-6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CT-3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant-fragment including the C-terminal 100 amino acids of human insulin receptor |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.