Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insulin Receptor (β-subunit) Ab-6 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS636R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1 mg/mL
200 μg/mL
Prediluted
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS636R7 7 mL Prediluted Prediluted
MS636P1 500 μL Concentrated 200 μg/mL
MS636P 1 mL Concentrated 200 μg/mL
MS636P0 100 μL Concentrated 200 μg/mL
MS636P1ABX 100 μL Concentrated 1 mg/mL
MS636PABX 200 μL Concentrated 1 mg/mL
MS636R1 1 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS636R7 Supplier Epredia Supplier No. MS636R7

Epredia™ Insulin Receptor (β-Subunit) Ab-6, Mouse Monoclonal Antibody provides accurate, reproducible results.

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain with tyrosine kinase acivity, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: CT-3

Isotype: IgG1

Species Reactivity: Human, Monkey, Mouse and Rat. Others not known.

Epitope: C-terminal

Immunogen: Recombinant fragment including the C-terminal 100 amino acids of human insulin receptor

Molecular Weight: 95kDa

Positive Control: HEP-G-2, IM-9 lymphocyte cells; Placenta or brain

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

  • Western Blotting
  • Immunoprecipitation (Not suitable)
  • Affinity Purification (With peptide elution)
  • ELISA (As capture antibody only)
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Insulin Receptor (beta-Subunit) Ab-6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CT-3
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant-fragment including the C-terminal 100 amino acids of human insulin receptor
Quantity 7 mL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human, Mouse, Non-human Primate, Rat
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.