Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Involucrin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide

Catalog No. MS126P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS126P 1 mL Unconjugated Concentrated 200μg/mL
MS126B0 100 μL Biotin Concentrated 200μg/mL
MS126P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS126PABX 200 μL Unconjugated Concentrated 1mg/mL
MS126B1 500 μL Biotin Concentrated 200μg/mL
MS126P1 500 μL Unconjugated Concentrated 200μg/mL
MS126B 1 mL Biotin Concentrated 200μg/mL
MS126R1 1 mL Unconjugated Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS126P Supplier Epredia™ Supplier No. MS126P

Epredia™ Involucrin Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Involucrin is expressed in a range of stratified squamous epithelia, including the cornea. In normal epidermis, it is first expressed in the upper spinous layers, and in keratinocyte cultures it is expressed by all cells that have left the basal layer. Involucrin expression is abnormal in squamous cell carcinomas and premalignant lesions, and is reduced in severe dysplasias of the larynx and cervix.

Host Species: Mouse

Clone: SY5

Isotype: IgG1

Species Reactivity: Human, Gorilla, Owl, Monkey, Pig, Dog and Rat. Others not known.

Epitope: Codon 421-568

Immunogen: Human keratinocytes' involucrin

Molecular Weight: 66-170kDa

Positive Control: LS174T cells, Skin

Cellular Localization: Cytoplasmic Ab-1 stains the involucrin in a variety of sizes: 170kDa in MCF-7 cells, a doublet of ∽115kDa and 150kDa in gorilla and owl monkey, 66kDa in dog, and a doublet of 105kDa in pig.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Involucrin Ab-1
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SY5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human keratinocytes involucrin
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cytoskeletal Protein
Primary or Secondary Primary
Target Species Canine, Human, Non-human Primate, Pig, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.