Learn More
Epredia™ Lab Vision™ Involucrin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: SY5
Isotype: IgG1
Species Reactivity: Human, Gorilla, Owl, Monkey, Pig, Dog and Rat. Others not known.
Epitope: Codon 421-568
Immunogen: Human keratinocytes' involucrin
Molecular Weight: 66-170kDa
Positive Control: LS174T cells, Skin
Cellular Localization: Cytoplasmic Ab-1 stains the involucrin in a variety of sizes: 170kDa in MCF-7 cells, a doublet of ∽115kDa and 150kDa in gorilla and owl monkey, 66kDa in dog, and a doublet of 105kDa in pig.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
- ELISA
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Involucrin Ab-1 |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | SY5 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human keratinocytes involucrin |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.