Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappa Light Chain Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS143P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS143P1 500 μL Unconjugated Concentrated 200μg/mL
MS143B0 100 μL Biotin Concentrated 200μg/mL
MS143B1 500 μL Biotin Concentrated 200μg/mL
MS143B 1 mL Biotin Concentrated 200μg/mL
MS143P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS143P 1 mL Unconjugated Concentrated 200μg/mL
MS143R7 7 mL Unconjugated Prediluted Prediluted
MS143P0 100 μL Unconjugated Concentrated 200μg/mL
MS143PABX 200 μL Unconjugated Concentrated 1mg/mL
MS143R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS143P1 Supplier Epredia Supplier No. MS143P1
Add to Cart
Add to Cart

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Kappa Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Antibody to the κ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Mouse

Clone: L1C1

Isotype: IgG1/k

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: B-lymphoma cells

Molecular Weight: 25kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Kappa Light Chain Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone L1C1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen B lymphoma cells
Quantity 500 μL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.