Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki-67 Ab-4 Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB1510P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
Purification Method:
Purified w/o BSA and Azide
Purified with BSA and Azide
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Purification Method
RB1510P1ABX 500 μL Concentrated Purified w/o BSA and Azide
RB1510P1 500 μL Concentrated Purified with BSA and Azide
RB1510P 1 mL Concentrated Purified with BSA and Azide
RB1510PABX 1 mL Concentrated Purified w/o BSA and Azide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB1510P1ABX Supplier Epredia Supplier No. RB1510P1ABX

Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67 Ab-4, Rabbit Polyclonal Antibody provides accurate, reproducible results. Rabbit Polyclonal Antibody.

Ki67 is a nuclear protein, which is expressed in the proliferating cells. Ki67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiscent) phase are negative for this protein.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: Middle of Ki-67 protein

Immunogen: A synthetic peptide from the human Ki-67 protein

Positive Control: Tonsil or Breast CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Ki-67 Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen A synthetic peptide from the human Ki67 protein
Purification Method Purified w/o BSA and Azide
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.