Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Rat. Others not known.
Epitope: Middle of Ki-67 protein
Immunogen: A synthetic peptide from the human Ki-67 protein
Positive Control: Tonsil or Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Ki-67 Ab-4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide from the human Ki67 protein |
| Purification Method | Purified w/o BSA and Azide |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.