Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ki-67 Rabbit Polyclonal Antibody, Epredia™

Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ Ki-67, Rabbit Polyclonal Antibody provides accurate, reproducible results.

Supplier:  Epredia RB9043R7

 View more versions of this product

Catalog No. RB9043R7


Description

Description

Ki-67 is a nuclear protein, which is expressed in the proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human Ki-67

Molecular Weight: 345kDa and 395kDa

Positive Control: Breast CA and lymph node

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Ki-67
Polyclonal
Rabbit
7 mL
Cancer and Tumor Biology
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
A synthetic peptide from C-terminus of human Ki-67
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.