Learn More
Description
Host Species: Mouse
Clone: N3H10
Isotype: IgG2b
Species Reactivity: Human, Monkey and Xenopus. Mouse and Hamster (Western Blotting only). Does not react with cow, rabbit and sea urchin.
Epitope: aa 506-541
Immunogen: Human placental extract designated as PSE1-PL
Molecular Weight: 70kDa
Positive Control: BT474, T47D, HEP-G-2 or HeLa cells; Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Ku (p70) Ab-4 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | N3H10 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human placental extract designated as PSE1-PL |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.