Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L1 Cell Adhesion Molecule Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS770P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS770P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS770B 1 mL Biotin Concentrated 200μg/mL
MS770R7 7 mL Unconjugated Prediluted Prediluted
MS770B0 100 μL Biotin Concentrated 200μg/mL
MS770B1 500 μL Biotin Concentrated 200μg/mL
MS770P0 100 μL Unconjugated Concentrated 200μg/mL
MS770PABX 200 μL Unconjugated Concentrated 1mg/mL
MS770P1 500 μL Unconjugated Concentrated 200μg/mL
MS770P 1 mL Unconjugated Concentrated 200μg/mL
MS770R1 1 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS770P1ABX Supplier Epredia Supplier No. MS770P1ABX
Add to Cart
Add to Cart

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting and Immunoprecipitation (Native and denatured), Epredia™ L1 Cell Adhesion Molecule Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

The L1CAM gene, which is located in Xq28, is involved in three distinct conditions: 1) HSAS (hydrocephalus-stenosis of the aqueduct of Sylvius); 2) MASA (mental retardation, aphasia, shuffling gait, adductus thumbs); and 3) SPG1 (spastic paraplegia). The L1, neural cell adhesion molecule (L1CAM) also plays an important role in axon growth, fasciculation, neural migration and in mediating neuronal differentiation. Expression of L1 protein is restricted to tissues arising from neuroectoderm.

Host Species: Mouse

Clone: UJ127

Isotype: IgG1/k

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Homogenous suspension of 16-week human fetal brain

Molecular Weight: 220-240kDa

Positive Control: IMR-5 cells Neuroblastoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen L1 Cell Adhesion Molecule Ab-1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone UJ127
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Homogenous suspension of 16 week human fetal brain
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.