Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Laminin Ab-1 Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB082R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB082R1 1 mL Prediluted
RB082A0 100 μL Concentrated
RB082A 1 mL Concentrated
RB082RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RB082R1 Supplier Epredia™ Supplier No. RB082R1

Recommended for Immunohistochemistry (Formalin/paraffin). Epredia™ Laminin Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Extracellular matrix (ECM) surrounds and separates most tissue compartments, determines tissue architecture and dynamically interacts with its surrounding cells. The basement membrane is a part of the ECM and is composed of collagens, proteoglycans, elastin and several glycoproteins of which laminin is the most abundant. Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains Dystroglycan is a major laminin receptor in muscle and epithelia, and nidogen is a high-affinity laminin-binding protein important for basement-membrane assembly.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Laminin isolated from a rat yolk sac tumor cell line

Positive Control: Skin, Lung, Kidney or Tonsil

Cellular Localization: Basement membrane Ab-1 labels basement membrane surrounding normal vessels, glands, muscles, nerves, fat and deciduas cells separating these from abutting connective tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Laminin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Laminin isolated from a rat yolk sac tumor cell line
Quantity 1 mL
Regulatory Status IVD
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.