Learn More
Description
Host Species: Rat
Clone: A5
Isotype: IgG2a/κ
Species Reactivity: Human and Mouse. Others not known.
Epitope: Not determined
Immunogen: Murine EHS laminin preparation
Molecular Weight: ∽210kDa
Positive Control: LS174T cells, Normal colon or colon CA
Cellular Localization: Basement membrane Specificity of Ab-1 was established by immunoprecipitation and immunofluorescence of human skeletal muscle and kidney with laminin chain-specific MAbs. Ab-1 stains uniformly all human and murine basement membranes.
Recommended for:
- Immunohistochemistry (Acetone-fixed frozen and acid-alcohol-fixed, paraffin-embedded)
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Laminin B2/gamma1 Ab-1 |
| Applications | Immunohistochemistry (Frozen), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | A5 |
| Concentration | 200µg/mL |
| Conjugate | Unconjugated |
| Host Species | Rat |
| Immunogen | Murine EHS laminin preparation |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.