Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin B2/γ1 Ab-1 Rat Monoclonal Antibody, Epredia™

Catalog No. RT795PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200µg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
RT795PABX 200 μL Unconjugated 1mg/mL
RT795B0 100 μL Biotin 200µg/mL
RT795B1 500 μL Biotin 200µg/mL
RT795B 1 mL Biotin 200µg/mL
RT795P0 100 μL Unconjugated 200µg/mL
RT795P 1 mL Unconjugated 200µg/mL
RT795P1ABX 100 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. RT795PABX Supplier Epredia Supplier No. RT795PABX
Add to Cart
Add to Cart

Recommended for Immunohistochemistry (Acetone-fixed frozen and acid-alcohol-fixed, paraffin-embedded), Immunofluorescence and Immunoprecipitation (Native and denatured), Epredia™ Laminin B2/γ1 Ab-1, Rat Monoclonal Antibody provides accurate, reproducible results.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of α, β, and γ chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.

Host Species: Rat

Clone: A5

Isotype: IgG2a/κ

Species Reactivity: Human and Mouse. Others not known.

Epitope: Not determined

Immunogen: Murine EHS laminin preparation

Molecular Weight: ∽210kDa

Positive Control: LS174T cells, Normal colon or colon CA

Cellular Localization: Basement membrane Specificity of Ab-1 was established by immunoprecipitation and immunofluorescence of human skeletal muscle and kidney with laminin chain-specific MAbs. Ab-1 stains uniformly all human and murine basement membranes.

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen and acid-alcohol-fixed, paraffin-embedded)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Laminin B2/gamma1 Ab-1
Applications Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone A5
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Rat
Immunogen Murine EHS laminin preparation
Quantity 200 μL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.