Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Receptor Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS259P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS259P1 500 μL 200μg/mL
MS259R7 7 mL -
MS259P0 100 μL 200μg/mL
MS259P1ABX 100 μL 1mg/mL
MS259PABX 200 μL 1mg/mL
MS259P 1 mL 200μg/mL
MS259R1 1 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS259P1 Supplier Epredia Supplier No. MS259P1
Add to Cart
Add to Cart

Recommended for Immunohistochemistry (Formalin/paraffin), Immunofluorescence and Flow Cytometry, Epredia™ Laminin Receptor Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Laminin, one of the major glycoproteins of basement membrane, displays multiple biological activities which are mediated through its interaction with specific cell membrane receptors. Overexpression of laminin receptor is reportedly an independent prognostic factor in breast cancer and correlates with the dissemination of the tumor cells in bone marrow. Laminin receptor appears to play an important role in tumor invasion and metastasis.

Host Species: Mouse

Clone: MLuC5

Isotype: IgM

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Small cell lung carcinoma N592 live cells

Molecular Weight: 67kDa

Positive Control: Breast CA

Cellular Localization: Membrane with some cytoplasmic Ab-1 partially inhibits the binding of soluble laminin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry
  • Inhibits (partially) the binding of soluble laminin

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Laminin Receptor Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MLuC5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Small cell lung carcinoma N592 live cells
Quantity 500 μL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgM
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.