Learn More
Description
p56lck is a membrane-associated non-receptor protein tyrosine kinase from the Src-family of kinases. The phosphorylation status and the activity of p56lck is regulated by the CD45 tyrosine protein phosphatase. p56lck plays a critical role in T cell development and activation. p56lck gene is localized to a site in the genome which undergoes frequent chromosomal abnormalities in lymphomas and neuroblastomas.
Host Species: Mouse
Clone: 3A5
Isotype: IgG2b/κ
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: aa 1-225
Immunogen: Recombinant protein corresponding to aa 1-225 of murine p56lck protein
Molecular Weight: 56kDa
Positive Control: Jurkat cells
Cellular Localization: Cell membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Denatured verified)
- Kinase Assay
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | LCK (p56superscriptlcksuperscript) Ab-1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 3A5 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to aa 1-225 of murine p56superscriptlcksuperscript protein |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.