Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCK (p56lck) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS164P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS164P0 100 μL 200μg/mL
MS164P1 500 μL 200μg/mL
MS164P 1 mL 200μg/mL
MS164P1ABX 100 μL 1mg/mL
MS164PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS164P0 Supplier Epredia Supplier No. MS164P0

Recommended for Western Blotting, Immunoprecipitation (Denatured verified) and Kinase Assays, Epredia™ LCK (p56lck) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

p56lck is a membrane-associated non-receptor protein tyrosine kinase from the Src-family of kinases. The phosphorylation status and the activity of p56lck is regulated by the CD45 tyrosine protein phosphatase. p56lck plays a critical role in T cell development and activation. p56lck gene is localized to a site in the genome which undergoes frequent chromosomal abnormalities in lymphomas and neuroblastomas.

Host Species: Mouse

Clone: 3A5

Isotype: IgG2b/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 1-225

Immunogen: Recombinant protein corresponding to aa 1-225 of murine p56lck protein

Molecular Weight: 56kDa

Positive Control: Jurkat cells

Cellular Localization: Cell membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen LCK (p56superscriptlcksuperscript) Ab-1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3A5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to aa 1-225 of murine p56superscriptlcksuperscript protein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.