Learn More
Description
Host Species: Mouse
Clone: LH01
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Purified human LH-β
Molecular Weight: 14kDa (β) and 28kDa (αβ)
Positive Control: Anterior pituitary
Cellular Localization: Cytoplasmic
Ab-1 recognizes the beta subunit both free and intact of luteinizing hormone (LH). Ab-1 is useful for the labeling of normal gonadotropic cells of the pituitary and also for the classification of pituitary adenomas and in the differential identification of primary and metastatic tumors of the pituitary.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | LH (Luteinizing Hormone) Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | LH01 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified human LH-Beta |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.