Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LH (Luteinizing Hormone) Ab-1, Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1448R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Form:
Concentrated
Prediluted, Ready-to-Use
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1448R1 1 mL Prediluted, Ready-to-Use
MS1448P0 100 μL Concentrated 200μg/mL
MS1448P1 500 μL Concentrated 200μg/mL
MS1448P1ABX 100 μL Concentrated 1mg/mL
MS1448PABX 200 μL Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1448R1 Supplier Epredia Supplier No. MS1448R1

Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ LH (Luteinizing Hormone) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Luteinizing hormone (LH) is a tropic hormone which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common α-subunit but also a spesofic β-subunit, which characterizes each of these hormones.

Host Species: Mouse

Clone: LH01

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human LH-β

Molecular Weight: 14kDa (β) and 28kDa (αβ)

Positive Control: Anterior pituitary

Cellular Localization: Cytoplasmic

Ab-1 recognizes the beta subunit both free and intact of luteinizing hormone (LH). Ab-1 is useful for the labeling of normal gonadotropic cells of the pituitary and also for the classification of pituitary adenomas and in the differential identification of primary and metastatic tumors of the pituitary.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen LH (Luteinizing Hormone) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LH01
Concentration
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human LH-Beta
Quantity 1 mL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Prediluted, Ready-to-Use
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.