Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ LH (Luteinizing Hormone) Mouse Monoclonal Antibody with BSA and azide

Catalog No. MS9078P Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS9078P 1 mL Concentrated
MS9078P1 500 μL Concentrated
MS9078R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS9078P Supplier Epredia™ Supplier No. MS9078P

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Ideal for Immunohistochemistry (Formalin/paraffin), Epredia™ LH (Luteinizing Hormone), Mouse Monoclonal Antibody provides accurate, reproducible results.

LH is a tropic hormone, which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common alpha-sububit but also a specific beta-subunit, which characterizes each of these hormones.

Host Species: Mouse

Clone: SPM103

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Immunogen: LH purified from human pituitary gland

Molecular Weight: 14kDa (β), 28kDa (α/β)

Positive Control: Anterior pituitary

Cellular Localization: Cytoplasmic. This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen LH (Luteinizing Hormone)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM103
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen LH purified from human pituitary gland
Quantity 1 mL
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.