Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP/MVP (Major Vault Protein) Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS664P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
Prediluted
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS664P 1 mL Unconjugated 200μg/mL
MS664P1ABX 100 μL Unconjugated 1mg/mL
MS664PABX 200 μL Unconjugated 1mg/mL
MS664P1 500 μL Unconjugated 200μg/mL
MS664R7 7 mL Unconjugated Prediluted
MS664B0 100 μL Biotin 200μg/mL
MS664B1 500 μL Biotin 200μg/mL
MS664B 1 mL Biotin 200μg/mL
MS664P0 100 μL Unconjugated 200μg/mL
MS664R1 1 mL Unconjugated Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MS664P Supplier Epredia Supplier No. MS664P
Add to Cart
Add to Cart

Recommended for a variety of techniques, Epredia™ LRP/MVP (Major Vault Protein) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Major vault protein (MVP) is identical to lung-resistance related protein (LRP). Its shape is reminiscent of the nucleopore central plug. Treatment of cells with estradiol increases the amount of MVP in nuclear extract. The hormone-dependent interaction of vaults with ER is prevented in vitro by sodium molybdate. Antibodies to progesterone and glucocorticoid receptors are also able to co-immunoprecipitate the MVP. Reportedly, the lung-resistance related protein (LRP) is identical to human MVP. LRP is overexpressed in many neoplastic tissues and cell lines indicating a poor response to chemotherapy.

Host Species: Mouse

Clone: 1032

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Proteins precipitated with AER317 from the phosphocellulose column flow through of extract of human breast cancer MCF-7 cells

Molecular Weight: 104-110kDa

Positive Control: SKBR-3 cells, Breast tumors

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Gel Supershift
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen LRP/MVP (Major Vault Protein) Ab-2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1032
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Proteins precipitated with AER317 from the phosphocellulose column flow through of extract of human breast cancer MCF-7 cells
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.