Learn More
Description
Host Species: Mouse
Clone: 1032
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Proteins precipitated with AER317 from the phosphocellulose column flow through of extract of human breast cancer MCF-7 cells
Molecular Weight: 104-110kDa
Positive Control: SKBR-3 cells, Breast tumors
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
- Gel Supershift
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | LRP/MVP (Major Vault Protein) Ab-2 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 1032 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Proteins precipitated with AER317 from the phosphocellulose column flow through of extract of human breast cancer MCF-7 cells |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.