Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MART-1/Melan-A (Melanoma Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS612P1ABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS612P1ABX 100 μL Concentrated 1mg/mL
MS612P0 100 μL Concentrated 200μg/mL
MS612P1 500 μL Concentrated 200μg/mL
MS612P 1 mL Concentrated 200μg/mL
MS612PABX 200 μL Concentrated 1mg/mL
MS612RQ 12 mL Ready-to-Use
MS612R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS612P1ABX Supplier Epredia Supplier No. MS612P1ABX
Add to Cart
Add to Cart

Epredia™ MART-1/Melan-A (Melanoma Marker) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.

Host Species: Mouse

Clone: M2-7C10

Isotype: IgG2b/κ

Species Reactivity: Human and Horse. Does not react with mouse and rat. Others not tested.

Epitope: Not determined

Immunogen: Recombinant hMART-1 protein

Molecular Weight: 18kDa

Positive Control: CAC1 melanoma cells and melanomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MART-1/Melan-A (Melanoma Marker) Ab-1
Applications Immunoprecipitation
Classification Monoclonal
Clone M2-7C10
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant hMART-1 protein
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Equine, Human
Form Concentrated
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.