Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody

Catalog No. MS799PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS799PABX 200 μL Concentrated 1mg/mL
MS799P0 100 μL Concentrated 200μg/mL
MS799P1 500 μL Concentrated 200μg/mL
MS799P1ABX 100 μL Concentrated 1mg/mL
MS799R7 7 mL Prediluted
MS799RQ 12 mL Ready-to-Use
MS799P 1 mL Concentrated 200μg/mL
MS799R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS799PABX Supplier Epredia™ Supplier No. MS799PABX
Add to Cart
Add to Cart

Epredia™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. MART-1 is present in melanosomes and endoplasmic reticulum.

Host Species: Mouse

Clone: A103

Isotype: IgG1

Species Reactivity: Human and Dog. Others not tested.

Epitope: Not determined

Immunogen: Recombinant MART-1 protein

Molecular Weight: 18kDa

Positive Control: CAC1 melanoma cells and Melanomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Specifications

Antigen MART-1/Melan-A (Melanoma Marker) Ab-4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A103
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant MART-1 protein
Quantity 200 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Canine, Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.