Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Mast Cell Chymase Ab-1 Mouse Monoclonal Antibody, 80μg/mL, BSA and azide

Catalog No. MS1217R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1217R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1217R1 Supplier Epredia™ Supplier No. MS1217R1

Epredia™ Mast Cell Chymase Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of chymase is estimated to be found in mast cells of skin and lung and suggestes this enzyme plays a major role in mast cell mediated events. Human mast cell chymase is considered to be an important marker of mast cells as well as an important mediator of inflammation.

Host Species: Mouse

Clone: CC1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human skin chymase

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Mast Cell Chymase Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CC1
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Purified human skin chymase
Quantity 1 mL
Regulatory Status RUO
Research Discipline Enzyme
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.