Learn More
Epredia™ Lab Vision™ Mast Cell Tryptase Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: AA1
Isotype: IgG1
Species Reactivity: Human, Monkey, Dog and Cat. Others not known.
Epitope: Not determined
Immunogen: Human mast cell tryptase purified from human lung tissues
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Mast Cell Tryptase Ab-2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | AA1 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human mast cell tryptase purified from human lung tissues |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.