Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Mast Cell Tryptase Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1216R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
50μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1216R1 1 mL Prediluted -
MS1216P1ABX 100 μL Concentrated 1mg/mL
MS1216P1 500 μL Concentrated 50μg/mL
MS1216R7 7 mL Prediluted -
MS1216PABX 200 μL Concentrated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1216R1 Supplier Epredia™ Supplier No. MS1216R1

Epredia™ Mast Cell Tryptase Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of tryptase is estimated to be found in mast cells of skin and lung and suggestes this enzyme plays a major role in mast cell mediated events. In vitro studies indicate tryptase can cleave C3 to form C3a anaphylatoxin, inactivate fibrinogen as a coaguable substrate for thrombin and activate latent collagenase. Human mast cell tryptase has considerable potential as a marker for mast cell activation as well as being a mediator of inflammation.

Host Species: Mouse

Clone: AA1

Isotype: IgG1

Species Reactivity: Human, Monkey, Dog and Cat. Others not known.

Epitope: Not determined

Immunogen: Human mast cell tryptase purified from human lung tissues

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Mast Cell Tryptase Ab-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone AA1
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Human mast cell tryptase purified from human lung tissues
Quantity 1 mL
Regulatory Status RUO
Research Discipline Enzyme
Primary or Secondary Primary
Target Species Canine, Feline, Human, Non-human Primate
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.