Learn More
Description
Host Species: Mouse
Clone: 5B2
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule
Molecular Weight: 40kDa
Positive Control: Mesothelioma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Mesothelin Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 5B2 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.