Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Mesothelin Ab-1 Mouse Monoclonal Antibody

Catalog No. MS1320S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1320S1 500 μL Concentrated
MS1320S 1 mL Concentrated
MS1320R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1320S1 Supplier Epredia™ Supplier No. MS1320S1
Add to Cart
Add to Cart

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mesothelin Ab-1, Mouse Monoclonal Antibody.

Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein, that is present on the surface of normal mesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignant mesotheliomas.

Host Species: Mouse

Clone: 5B2

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule

Molecular Weight: 40kDa

Positive Control: Mesothelioma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Mesothelin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5B2
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to the membrane-bound form of the mesothelin molecule
Quantity 500 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.