Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Mesothelioma Ab-1 Mouse Monoclonal Antibody

Catalog No. MS1494R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
12 mL
Form:
Prediluted
Ready-to-Use
Supernatant
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1494R1 1 mL Prediluted
MS1494R7 7 mL Prediluted
MS1494S1 500 μL Supernatant
MS1494S 1 mL Supernatant
MS1494RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1494R1 Supplier Epredia™ Supplier No. MS1494R1

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Epredia™ Mesothelioma Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: HBME-1

Isotype: IgM/κ

Species Reactivity: Human. Others not known.

Immunogen: A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma

Positive Control: Mesothelioma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Mesothelioma Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HBME-1
Conjugate Unconjugated
Host Species Mouse
Immunogen A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.