Learn More
Description
Host Species: Mouse
Clone: HBME-1
Isotype: IgM/κ
Species Reactivity: Human. Others not known.
Immunogen: A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma
Positive Control: Mesothelioma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Mesothelioma Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | HBME-1 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A suspension of human mesotheliema cells from patients with malignant epithelial mesothelioma |
| Quantity | 7 mL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.