Learn More
Description
Host Species: Mouse
Clone: 246-E8.E7; same as HLA-ABC-m2
Isotype: IgG2a
Species Reactivity: Human. Does not react with primate cells. Others not tested.
Epitope: Not determined
Immunogen: Human PBL (T-ALL patient)
Molecular Weight: 12kDa
Positive Control: Jurkat cells
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Microglobulin beta-2 Ab-1 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 246-E8.E7 ;same as HLA-ABC-m2 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human PBL (T-ALL patient) |
| Purification Method | Jurkat cells |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.