Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microglobulin β -2 Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS440PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS440PABX 200 μL 1mg/mL
MS440P0 100 μL 200μg/mL
MS440P1 500 μL 200μg/mL
MS440P 1 mL 200μg/mL
MS440P1ABX 100 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS440PABX Supplier Epredia Supplier No. MS440PABX

Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments with the Epredia™ Microglobulin β-2 Ab-1, Mouse Monoclonal Antibody.

Β2-microglobulin non-covalently associates with the 44 kDa α chain to form the HLA Class I antigen complex. Human Β2-microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. Clone HLA-ABC-m2 (also known as 246-E8.E7) is specific to human Β-microglobulin and does not react with primate cells.

Host Species: Mouse

Clone: 246-E8.E7; same as HLA-ABC-m2

Isotype: IgG2a

Species Reactivity: Human. Does not react with primate cells. Others not tested.

Epitope: Not determined

Immunogen: Human PBL (T-ALL patient)

Molecular Weight: 12kDa

Positive Control: Jurkat cells

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Microglobulin beta-2 Ab-1
Applications Immunofluorescence
Classification Monoclonal
Clone 246-E8.E7 ;same as HLA-ABC-m2
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human PBL (T-ALL patient)
Purification Method Jurkat cells
Quantity 200 μL
Regulatory Status RUO
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.