Learn More
Microphthalmia (Transcription Factor) (MiTF) Ab-2 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: D5
Isotype: IgG1/κ
Species Reactivity: Human and Dog. Does not react with mouse and rat. Others not known.
Epitope: N-terminal
Immunogen: N-terminal fragment of human Mi protein
Molecular Weight: 52-56kDa doublet
Positive Control: 501 Mel cells, Melanoma
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not verified)
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Microphthalmia (Transcription Factor) (MiTF) Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | D5 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | N-terminal fragment of human Mi protein |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.