Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mitochondria Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1372P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS1372P1 500 μL Unconjugated Concentrated 200μg/mL
MS1372P 1 mL Unconjugated Concentrated 200μg/mL
MS1372B0 100 μL Biotin Concentrated 200μg/mL
MS1372B1 500 μL Biotin Concentrated 200μg/mL
MS1372P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS1372PABX 200 μL Unconjugated Concentrated 1mg/mL
MS1372R1 1 mL Unconjugated Prediluted -
MS1372R7 7 mL Unconjugated Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS1372P1 Supplier Epredia Supplier No. MS1372P1
Add to Cart
Add to Cart

Epredia™ Mitochondria Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Host Species: Mouse

Clone: MTC02

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Semi-purified mitochondrial preparation

Molecular Weight: 60kDa

Positive Control: HeLa cells, Liver

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Mitochondria Ab-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MTC02
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Semi-purified mitochondrial preparation
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.