Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ MMP-11 (Stromelysin-3) Ab-5 Mouse Monoclonal Antibody, 200μg/mL, biotin labeled with BSA and azide

Catalog No. MS1035P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS1035P0 100 μL Unconjugated Concentrated 200μg/mL
MS1035B0 100 μL Biotin Concentrated 200μg/mL
MS1035P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS1035PABX 200 μL Unconjugated Concentrated 1mg/mL
MS1035P1 500 μL Unconjugated Concentrated 200μg/mL
MS1035B 1 mL Biotin Concentrated 200μg/mL
MS1035P 1 mL Unconjugated Concentrated 200μg/mL
MS1035R1 1 mL Unconjugated Prediluted -
MS1035R7 7 mL Unconjugated Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS1035P0 Supplier Epredia™ Supplier No. MS1035P0

Ensure accurate, reproducible results in experiments with the Epredia™ MMP-11 (Stromelysin-3) Ab-5, Mouse Monoclonal Antibody.

Stromelysin, a member of the matrix metalloproteinase family, demonstrates wide substrate specificity with the ability to degrade proteoglycan, fibronectin, laminin, casein, and the nonhelical region of collagen. The stromelysin-3 (MMP-11) gene was originally identified on the basis that it is expressed specifically in stromal cells surrounding invasive breast carcinomas. MMP-11 is localized by in situ hybridization to the long arm of chromosome 22. The MMP-11 expression is found more specifically in malignant tumors than in benign ones.

Host Species: Mouse

Clone: SL3.05

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Immunogen: Recombinant protein corresponding to active form of human MMP-11

Molecular Weight: ∽60kDa (latent), ∽47kDa (active) and ∽35kDa (degradation product)

Positive Control: Recombinant MMP-11 protein; Placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MMP-11 (Stromelysin-3) Ab-5
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SL3.05
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to active form of human MMP-11
Quantity 100 μL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.