Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-2 (72kDa Collagenase IV) Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS804B0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS804B0 100 μL Biotin 200μg/mL
MS804B1 500 μL Biotin 200μg/mL
MS804B 1 mL Biotin 200μg/mL
MS804P0 100 μL Unconjugated 200μg/mL
MS804P1 500 μL Unconjugated 200μg/mL
MS804P 1 mL Unconjugated 200μg/mL
MS804P1ABX 100 μL Unconjugated 1mg/mL
MS804PABX 200 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS804B0 Supplier Epredia Supplier No. MS804B0

Epredia™ MMP-2 (72kDa Collagenase IV) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids.

Host Species: Mouse

Clone: VB3

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human native 72kDa MMP-2

Molecular Weight: 72kDa (proform) and ∽66kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MMP-2 (72kDa Collagenase IV) Ab-2
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone VB3
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Human native 72kDa MMP-2
Quantity 100 μL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.