Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-3 (Stromelysin-1) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS809B1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Conjugate:
Biotin
Unconjugated
Concentration:
1mg/mL
200μg/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Concentration
MS809B1 500 μL Biotin 200μg/mL
MS809B0 100 μL Biotin 200μg/mL
MS809B 1 mL Biotin 200μg/mL
MS809P0 100 μL Unconjugated 200μg/mL
MS809P1 500 μL Unconjugated 200μg/mL
MS809P 1 mL Unconjugated 200μg/mL
MS809P1ABX 100 μL Unconjugated 1mg/mL
MS809PABX 200 μL Unconjugated 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS809B1 Supplier Epredia Supplier No. MS809B1

Epredia™ MMP-3 (Stromelysin-1) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Matrix metalloproteinases (MMP) are proteolytic enzymes capable of degrading connective tissue components. Imbalanced secretion of certain MMP or disturbances in the differential control of MMP by tissue inhibitor of MMPs (TIMPs) are implicated in the invasive potential of malignant tumors. Stromelysin (MMP3/Transin) degrades several matrix components including the core proteins of proteoglycans, laminin and non-helical regions of collagens.

Host Species: Mouse

Clone: SL-1 ID3

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)

Molecular Weight: 54/59kDa (proform) and 44/49kDa (active form)

Positive Control: Conditioned, serum-free medium from TPA-treated human endometrial cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not Suitable)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MMP-3 (Stromelysin-1) Ab-1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone SL-1 ID3
Concentration 200μg/mL
Conjugate Biotin
Host Species Mouse
Immunogen APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)
Quantity 500 μL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.