Learn More
Description
Host Species: Mouse
Clone: SL-1 ID3
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1)
Molecular Weight: 54/59kDa (proform) and 44/49kDa (active form)
Positive Control: Conditioned, serum-free medium from TPA-treated human endometrial cells
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Not Suitable)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | MMP-3 (Stromelysin-1) Ab-1 |
| Applications | Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | SL-1 ID3 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | APMA (4-Aminophenylmercuric acetate) activated Human stromelysin-1 (SL-1) |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.