Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ MMP-9 (92kDa Collagenase IV) Rabbit Polyclonal Antibody with BSA and azide

Catalog No. RB9234R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB9234R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RB9234R1 Supplier Epredia™ Supplier No. RB9234R1

Epredia™ MMP-9 (92kDa Collagenase IV), Rabbit Polyclonal Antibody provides accurate, reproducible results.

Metallomatrix proteases (MMPs) are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. This antibody reacts with both latent and active MMP-9.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Guinea Pig. Does not react with mouse, rat or bovine. Others not tested.

Epitope: Near C-terminus

Immunogen: A synthetic peptide derived from near C-terminal of human MMP-9 protein

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Placenta, or Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MMP-9 (92kDa Collagenase IV)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Rabbit
Immunogen A synthetic peptide derived from near C-terminal of human MMP-9 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Guinea Pig
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.