Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC-1 Ab-5 Hamster Monoclonal Antibody, Epredia™

Catalog No. HM1630R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
HM1630R1 1 mL Prediluted
HM1630P1 500 μL Concentrated
HM1630R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. HM1630R1 Supplier Epredia Supplier No. HM1630R1

Epredia™ MUC-1 Ab-5, Hamster Monoclonal Antibody provides accurate, reproducible results.

Muc1 is a heavily O-glycosylated transmembrane protein expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed in >90% breast carcinomas and metastases. In normal mammary gland it is expressed in apical surface of glandular epithelium. In breast cancer Muc1 is overexpressed; is underglycosylated and the apical localization is lost. Muc1 is transcribed as a larger precursor which is cleaved to form a larger mucin like subunit (265-400kDa) and a smaller subunit (14-28kDa) noncovalently associated with each other. Transgenic Muc1 has been shown to associate with all four erbB receptors and localize with erbB1 (EGFR) in lactating glands1. Muc1 can act as a ligand for ICAM-1 on HUVEC cells; it can bind beta-catenin, GSK3beta and it associates with Grb2-SOS upon phosphorylation.

Host Species: Hamster

Clone: MH1 (CT2)

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: aa 239-255

Immunogen: A synthetic peptide corresponding to aa 239-255 (SSLSYTNPAVAATSANL) from the cytoplasmic tail of MUC-1

Molecular Weight: 14-28kDa

Positive Control: MCF-7 cells; Breast CA

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MUC-1 Ab-5
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MH1; same as CT2
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Hamster
Immunogen A synthetic peptide corresponding to aa 239-255 (SSLSYTNPAVAATSANL) form the cytoplasmic tail of MUC1
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Mouse
Form Prediluted
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.