Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ MUC-1 Mouse Monoclonal Antibody, 200μg/mL with BSA and azide

Catalog No. MS10340R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS10340R7 7 mL Prediluted
MS10340P0 100 μL Concentrated
MS10340P1 500 μL Concentrated
MS10340P 1 mL Concentrated
MS10340R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS10340R7 Supplier Epredia™ Supplier No. MS10340R7

Deliver accurate, reproducible results with the Epredia™ MUC-1, Mouse Monoclonal Antibody.

MUC-1 is a heavily O-glycosylated transmembrane protein expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC-1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands.

Host Species: Mouse

Clone: SPM492

Isotype: IgG1/k

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Human breast cell line ZR-75-1

Molecular Weight: 265-400kDa

Positive Control: Breast CA

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MUC-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM492
Conjugate Unconjugated
Host Species Mouse
Immunogen Human breast cell line ZR-75-1
Purification Method Breast CA
Quantity 7 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.