Learn More
Description
Host Species: Mouse
Clone: M53
Isotype: IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein encoding five tandem repeats of the human MUC2 apomucin
Molecular Weight: 520kDa
Positive Control: Small intestine
Cellular Localization: Cytoplasmic and cell surface
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Mucin 2 (MUC2) Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | M53 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein encoding 5 tandem repeats of the human MUC2 apomucin |
| Quantity | 200 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.