Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucin 2 (MUC2) Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1037R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1037R1 1 mL Prediluted
MS1037P1 500 μL Concentrated 200μg/mL
MS1037P 1 mL Concentrated 200μg/mL
MS1037P1ABX 100 μL Concentrated 1mg/mL
MS1037PABX 200 μL Concentrated 1mg/mL
MS1037R7 7 mL Prediluted
MS1037RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1037R1 Supplier Epredia Supplier No. MS1037R1

Deliver accurate, reproducible results with the Epredia™ Mucin 2 (MUC2) Ab-2, Mouse Monoclonal Antibody.

Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.

Host Species: Mouse

Clone: M53

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein encoding five tandem repeats of the human MUC2 apomucin

Molecular Weight: 520kDa

Positive Control: Small intestine

Cellular Localization: Cytoplasmic and cell surface

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Mucin 2 (MUC2) Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone M53
Concentration
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Recombinant protein encoding 5 tandem repeats of the human MUC2 apomucin
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.