Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucin 3 (MUC3) Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1734P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1734P 1 mL 200μg/mL
MS1734P0 100 μL 200μg/mL
MS1734P1 500 μL 200μg/mL
MS1734P1ABX 100 μL 1mg/mL
MS1734PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1734P Supplier Epredia Supplier No. MS1734P

Epredia™ Mucin 3 (MUC3) Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Mucin 3 (MUC3) is a glycoprotein. Distributed in colon and rectum, and to a lesser extent in breast, lung and salivary gland tissues.

Host Species: Mouse

Clone: 1143/B7

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Tandem repeats of the human MUC3

Immunogen: MUC3 tandem repeat peptide

Molecular Weight: High

Positive Control: Colon CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Mucin 3 (MUC3) Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1143/B7
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen MUC3 tandem repeat peptide
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.