Learn More
Description
Host Species: Mouse
Clone: 1143/B7
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Epitope: Tandem repeats of the human MUC3
Immunogen: MUC3 tandem repeat peptide
Molecular Weight: High
Positive Control: Colon CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Mucin 3 (MUC3) Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 1143/B7 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | MUC3 tandem repeat peptide |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.