Learn More
Description
Host Species: Mouse
Clone: 45M1
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Rabbit, Cat, Mouse, Rat, Pig, Hedgehog and Chicken. Does not react with cow. Others not known.
Epitope: Destroyed by β-mercaptoethanol and proteases but not by periodate treatment
Immunogen: M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient
Molecular Weight: >1000kDa
Positive Control: Stomach
Cellular Localization: Cytoplasmic and cell surface
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 45M1 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.