Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS145R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS145R7 7 mL Unconjugated Prediluted Prediluted
MS145B 1 mL Biotin Concentrated 200μg/mL
MS145P1 500 μL Unconjugated Concentrated 200μg/mL
MS145B1 500 μL Biotin Concentrated 200μg/mL
MS145P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS145P 1 mL Unconjugated Concentrated 200μg/mL
MS145R1 1 mL Unconjugated Prediluted Prediluted
MS145RQ 12 mL Unconjugated Ready-to-Use Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MS145R7 Supplier Epredia Supplier No. MS145R7

Deliver accurate, reproducible results with the Epredia™ Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1, Mouse Monoclonal Antibody.

Mucins are high molecular eight glycoproteins with 80% carbohydrate contents and the remaining 20% is constituted by protein core. Gastric mucin is found in normal human gastrointestinal tract. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.

Host Species: Mouse

Clone: 45M1

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Rabbit, Cat, Mouse, Rat, Pig, Hedgehog and Chicken. Does not react with cow. Others not known.

Epitope: Destroyed by β-mercaptoethanol and proteases but not by periodate treatment

Immunogen: M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient

Molecular Weight: >1000kDa

Positive Control: Stomach

Cellular Localization: Cytoplasmic and cell surface

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 45M1
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient
Quantity 7 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Chicken, Feline, Hedgehog, Human, Mouse, Non-human Primate, Pig, Rabbit, Rat
Form Prediluted
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.