Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myeloperoxidase (MPO) Rabbit Polyclonal Antibody, Epredia™

Catalog No. RB373A0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RB373A0 100 μL Concentrated
RB373A1 500 μL Concentrated
RB373A 1 mL Concentrated
RB373R7 7 mL Ready-to-Use
RB373RQ 12 mL Ready-to-Use
RB373R1 1 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. RB373A0 Supplier Epredia Supplier No. RB373A0
Add to Cart
Add to Cart

Myeloperoxidase (MPO), Rabbit Polyclonal Ab is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Myeloperoxidase is an important enzyme used by granulocytes during phagocytic lysis of foreign particles engulfed. In normal tissues and in a variety of myeloproliferative disorders myeloid cells of both neutrophilic and eosinophilic types, at all stages of maturation, exhibit strong cytoplasmic reactivity for MPO. Erythroid precursors, megakaryocytes, lymphoid cells, mast cells, and plasma cells are nonreactive. MPO is not observed in the neoplastic cells of a wide variety of epithelial tumors and sarcomas. MPO is useful in differentiating between myeloid and lymphoid leukemias.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human granulocytic MPO

Positive Control: Tonsil or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Myeloperoxidase (MPO)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen Purified human granulocytic MPO
Quantity 100 μL
Regulatory Status IVD
Research Discipline Enzyme
Primary or Secondary Primary
Target Species Human, Mouse
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.