Learn More
Epredia™ Lab Vision™ Myoglobin Rabbit Polyclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Purified human myoglobin
Molecular Weight: 17.8kDa
Positive Control: Skeletal muscle
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Myoglobin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 200µg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Purified human myoglobin |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Research Discipline | Muscle Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.